Zinc Sulfate 7-Hydrate

Supplier: ALDON CORP SE

ZZ0110-500G ZZ0110-100G ZZ0110-500GR
470303-142EA 35.55 CAD
470303-142 470303-140 470303-144
Zinc Sulfate 7-Hydrate
Chemicals for Science Education

CAS Number: 7446-20-0
Formula Weight: 287.54
Formula: ZnSO4·7H2O
Hazard Info: Irritant
Density (g/mL): 1.97
Freezing Point (°C): 100
Solubility: Water
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR