Zinc Chloride

Supplier: ALDON CORP SE

ZZ0060-500G ZZ0070-500ML ZZ0069-CONC ZZ0060-100G
470303-080EA 52.2 CAD
470303-080 470303-074 470303-076 470303-078
Zinc Chloride
Chemicals for Science Education

CAS Number: 7646-85-7
Formula: ZnCl2
Density: 2.91 g/mL
Boiling Point: 732°C
Freezing Point: 290°C
Solubility: Water and Alcohol
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is corrosive.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR