Zinc

Supplier: ALDON CORP SE

ZZ0038-1SH ZZ0021-500G ZZ0021-100GR ZZ0015-2.5KG ZZ0015-500G ZZ0023-25G ZZ0030-500G ZZ0030-500GR ZZ0037-1SHT ZZ0026-500G ZZ0022-100G ZZ0022-500G ZZ0022-500GR ZZ0035-100G ZZ0035-500GR
470104-622EA 44.25 CAD
470104-622 470045-544 470303-084 470303-088 470303-090 470303-094 470303-096 470303-098 470303-102 470303-082 470303-106 470303-108 470303-110 470303-112 470303-114
Zinc
Chemicals for Science Education

CAS Number: 7440-66-6
Formula: Zn
Density: 7.12 g/mL
Boiling and Freezing Point: 907°C, 419°C
Solubility: Acids and Alkalies
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR