Yeast

Supplier: ALDON CORP SE

YY0005-100G YY0005-500G YY0010-100G YY0010-500G
470303-060EA 29.3 CAD
470303-060 470303-062 470303-064 470303-066
Yeast
Chemicals for Science Education

CAS Number: 6876-77-7 Brewer's/8013-01-2 Active Dry
Formula: Mixture
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR