Wright’s Blood Stain

Supplier: ALDON CORP SE

WW0060-100ML WW0060-500ML
470303-050EA 17.05 CAD
470303-050 470303-052
Wright’s Blood Stain
Chemicals for Science Education

CAS Number: 68988-92-1
Hazard Info: Flammable
Density (g/mL): 0.79
Boiling Point (°C): 65
Freezing Point (°C): -98
Solubility: Alcohol and water
Shelf Life (months): 36
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR