Wooden Meter and Half-Meter Sticks
Supplier: AMERICAN SCIENTIFIC LLC SE
2192-98 104905
470017-002EA 10.25 CAD
470017-002 470005-628
Wooden Meter and Half-Meter Sticks
Rulers
Good quality hardwood meter and half-meter sticks with a natural finish.
  • Permanently engraved scales
  • One side is graduated in cm with 1mm divisions
  • Other side is graduated in inches with 1/8-inch divisions
Meter sticks are 6mm thick x 25.4mm wide.