Wide-Mouth Polypropylene Cylinders
Supplier: SI Manufacturing
78130 78115 78110 78120-A 78125-A
470020-852EA 22.45 CAD
470020-852 470150-800 470150-424 470020-848 470020-850
Wide-Mouth Polypropylene Cylinders
Cylinders Measuring Cylinders
Chemical and Heat Resistant to 80°C.
  • Wide mouth is ideal for performing density tests
  • Molded, blue over-printed graduations
  • Aligned with GLOBE protocols
The squat design increases stability on the bench while the low form requires less overhead storage space.