Whose DNA Was Left Behind?
Supplier: Edvotek
S-51
470200-480EA 143.65 CAD
470200-480
Whose DNA Was Left Behind?
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
Students identify individuals based on DNA analysis.
  • Convenient, Ready-To-Load Samples
  • Time required: 45 minutes
  • Students Identify Individuals Based on DNA Analysis
  • Electrophoresis Buffer Included
DNA obtained from a single hair left behind at a crime scene can be used to identify a criminal.
In this 45 minute experiment, students compare simulated crime scene DNA with that of two suspects.
Ordering information: This kit includes Ready-to-Load dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes. Electrophoresis apparatus, a power supply, and a microwave or hot plate are required but not included.
Delivery information: Store Ready-to-Load QuickStrip samples in the refrigerator after they are delivered.
Packaging: Store Ready-to-Load QuickStrip™ samples in the refrigerator.