Ward's® Live White Planaria (Procotyla fluviatilis)
Supplier: Avantor
470175-822PK 47.4 CAD
470175-822 470180-238 470180-240
Ward's® Live White Planaria (Procotyla fluviatilis)
Specimens Live Specimens Animals - Living Material
Planaria are a flatworm commonly used in science fair projects. They do not bite and move very slowly, making them easy for young students to observe.
  • Study structure, feeding habits, responses to environmental stimuli and regeneration
  • Compare to Brown and White Planaria (also available)
  • Multiple colored specimens in one sample allowing side-by-side comparison
  • Vital Stained cultures allow students to better visualize organism structures
  • Click on "Product Resources" tab for additional materials for this product
Though smaller than other types ofMore Product Information