Ward's® What Influences Enzyme Activity? Kit
Supplier: Avantor
470015-314KT 127 CAD
470015-314 470014-612
Ward's® What Influences Enzyme Activity? Kit
Educational Classroom Kits and Activities
This kit demonstrates the process of starch digestion.
  • Lab Teaches Students to Recognize Breakdown of Starch into Glucose
  • Four Activities in One Kit
  • Pre and Post-Laboratory Assessments Included
  • Estimated Lab Time: 30 to 40 minutes
  • Material for 15 Setups
  • Aligned to NGSS standards (MS/HS: PS1, PS2, LS1)
This lab uses four lab activities and the enzyme diastase of malt to explore the breakdown of starch into glucose. Students perform a starch test and observe the breakdown of starch into glucose to learn how concentration, temperature, and pH affect theMore Product Information