Welch Vacuum Pumps
Supplier: Welch Vacuum
2511B-01 2534B-01
470120-990EA 888.75 CAD
470120-990 470120-992
Welch Vacuum Pumps
Pumps Vacuum Pumps
Welch Vacuum Pumps are perfect for vacuum or pressure applications.
  • Oil-Free Operation
  • Compact
  • Economical
  • Ideal Purposes Include Desiccating, Aspiration, Degassing, and More
These compact, economical pumps operate oil-free.
They're best suited for vacuum drying, desiccating, aspiration, degassing, chamber roughing, and filtration.