Water Still
Supplier: Dynalon
4000-0000 540004-0120
470003-602EA 4366.15 CAD
470316-884 470003-602
Water Still
Water Purification Systems Stills
Two capacities available; 2.4L/hr.and 4.0L/hr.
  • Complies to ASTM and ISO standards
  • Pyrogen free single distilled water
  • Easy to clean – no glass dismantling
  • Bulit-in condenser to cool water
Still features a thermostat that protects it in the event of a water supply failure and automatically resets when the still cools. It’s easy to clean by adding cleaning agents through a built-in funnel. It also comes with a drainage stopcock and metal stand for mounting to a wall or tabletop.
Ordering information: Available to U.S. customers only.