Watchmaker Forceps
Supplier: DUNRITE INSTRUMENTS SE
140505 141400
470126-100EA 11.65 CAD
470126-100 470018-868
Watchmaker Forceps
Forceps
Ultra fine control.
  • Super fine point for precise work
  • Smooth surface prevents damage to delicate tissues
  • Broad handle for easy control
This is the traditional instrument for fine dissection work, with fine points and a broad handle. Choose between the standard broad handled, and economy narrow handled versions, both with very sharp, smooth tips.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.