Ward's® WARDSafe™
Supplier: ALDON CORP SE
WW0095-3.8L WW0095-19L
470233-544EA 134.75 CAD
470233-544 470233-546
Ward's® WARDSafe™
Chemicals for Science Education
Specimen Holding Fluid, Dilute one part to nine parts water for use.
Shelf Life (months): 36
Storage: Red
  • Shelf Life (months): Indefinite
  • Storage: Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.