Wall Service Bench, Phenolic Resin Surface
Supplier: Diversified Woodcrafts
3214K 3224K 3234K 3244K
470117-722EA 10391.2 CAD
470117-722 470117-724 470117-726 470117-728
Wall Service Bench, Phenolic Resin Surface
Furniture Tables and Workbenches
Need all your services in one place or looking to add a wall of cabinets that include services? The Wall Service Bench offers storage and services all in one unit.
  • Constructed of solid oak and oak veneers
  • Outer ends offer a locking cabinet with one full height door that includes one adjustable shelf and four locking drawers
The Wall Service Bench features your choice of a 1" solid epoxy resin or ¾" phenolic resin top with an epoxy resin sink and two combination cold water/gas fixtures. The sink area allows access to the plumbing and provides a large storage space with two GFI protected AC duplex electrical outlets. There is enough room in the sink area to allow two 24" D tables to be butted against the unit for use as a peninsula work area. The outer ends of the unit offer four storage solutions: doors with adjustable shelves, doors/drawer combination with one adjustable shelf, four drawers/doors with one adjustable shelf and four drawers. All drawers and doors have locks and ADA compliant pulls. The unit is finished with a chemical resistant, earth-friendly UV finish and includes rubber base molding.
Sink dimensions: 18L×15W×11"D
Certifications: SEFA compliant and MAS Certified Green.