VWR® Volumetric Flasks
Supplier: VWR International
470148-692 470148-798 470148-806 470148-908 470149-060 470149-064
470148-692CS 606.1 CAD
470148-692 470148-798 470148-806 470148-908 470149-060 470149-064
VWR® Volumetric Flasks
Flasks Volumetric Flasks
Our borosilicate flasks meet ASTM standards.
  • Class A Flasks
  • Borosilicate Glass Construction
  • Graduated Marking for Measurements
  • Economical
  • Snap-Caps Included
The necks fit the included snap-caps.
The Class A, volumetric flasks include a graduation line for accurate measurements.