VWR® Micropipette Tip Reload System
Supplier: Thermo Scientific
112NXL T113RN-V T113RLN-V 104 T104R-V T112NXLRL-V 113-GN 104 T113RN-V 113-GN 112NXL
470146-156CS 1021.3 CAD
470146-156 470146-178 470146-212 470146-222 470146-224 470146-242 470146-264 470146-722 470200-716 470200-718 470200-720 470200-724
VWR® Micropipette Tip Reload System
Pipette Tips
Save time, plastic, and money with the Micropipette tip reload system.
  • Tips available in three different configuarations
  • Save plastic and re-use VWR® hinged rack boxes with the Reload Sleeves
  • Flip-lock boxes offer easy one-handed operation
  • Lid lip enables stable stacking
  • Tips work with most manufacturer's micropipettors
The reloading operation features sleeve alignment, which orients loading for a perfect fit every time.
The tongue and groove format keeps the insert fastened while applying tips, and the tall clip makes the insert easy to handle without touching tips with your hand.
Packaging: The Racked Tips contain 10 boxes ('racks') of 96 per pack. There are 5 packs per case.
The Reload Sleeves contain 10 inserts of 96 tips per pack. There are 5 packs per case.
The Bulk Bags contain 1000 loose tips per bag. There are 10 bags per case.