Single-Scale Precision Cylinder
Supplier: VWR International
470013-536 470148-656 470148-788 470148-844 470148-742
470013-536EA 213.75 CAD
470013-536 470148-656 470148-788 470148-844 470148-742
Single-Scale Precision Cylinder
Cylinders Measuring Cylinders
Our high-quality borosilicate cylinders meet ASTM standards.
  • Cylinders Calibrated to Contain
  • Borosilicate Cylinders
  • Uniform Wall Thickness
  • Molded Base
  • Graduated Metric Scale
Cylinders feature uniform wall thickness, white enamel, a single and graduated metric scale, pour-out, and a molded base.
They can be used with acids, salt solutions, and organic solvents.
Order Now
SPECIFICATIONS
Form Tall
Environmentally Preferable