VWR® Measuring Pipettes
Supplier: VWR International
470149-276 470149-278 470149-280 470149-282
470149-276CS 498.65 CAD
470149-276 470149-278 470149-280 470149-282
VWR® Measuring Pipettes
Pipettes Measuring (Mohr) Pipettes
These high-quality borosilicate, Mohr-type pipettes meet ASTM standards.
  • Color-coded by size
  • Calibrated 'to deliver'
  • Measurements are accurate to ±0.01 ml
The pipettes feature colored graduations in 0.1 increments that are enameled into the glass to prevent fading.