VWR® Low Form Heavy Duty Beakers
Supplier: VWR International
470148-638 470190-850 470191-232 470191-234 470191-236 470191-238
470148-638CS 353.3 CAD
470148-638 470190-850 470191-232 470191-234 470191-236 470191-238
VWR® Low Form Heavy Duty Beakers
Beakers
Our high-quality borosilicate beakers meet ASTM standards at a great value.
  • Borosilicate glass fabrication
  • Double-graduated metric scale
They feature a 0.5 mm thicker wall than standard-grade beakers; a white enamel, double-graduated metric scale; and a marking spot. Each can be heated and used with acids, salt solutions, and organic solvents.