Ward's Compact Scales
Supplier: VWR International
470003-174 30467773
470003-174EA 270.6 CAD
470003-174 470314-390
Ward's Compact Scales
Balances and Scales
These portable, compact scales are an economic option for the lab.
  • Auto shut-off
  • One-year manufacturer's warranty
  • High-contrast LCD Screen
  • Two-button keypad
  • Stackable
  • Aligned with GLOBE protocols
These models feature a simple two-button keypad and high-contrast LCD screen for ease of use and visibility.
Accessories information: An AC adapter is available separately.
Ordering information: One-year manufacturer's warranty is included with purchase. The scales include three AA batteries.