Variable Speed Electrostatic Generator
Supplier: Science First
RP-601 N-100E
470005-528EA 43.2 CAD
470005-528 470005-526
Variable Speed Electrostatic Generator
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This generator can perform all electrostatic experiments on a spectacular scale.
  • Rugged Construction
  • Troubleshooting Guide Included
  • Generator Produces 250,000 to 400,000 Volts
  • Polished, Smudge-Resistant Finish
  • Extra Belt Included
This variable speed Van de Graaff-type generator produces 250,000 to 400,000 volts. It's built to give maintenance-free operation for many years. The sphere is 25 cm in diameter with a polished, smudge-resistant finish. The column between the base and sphere is made of clear Lucite.
The overall height of the unit is 76 cm.More Product Information