Variable Speed Centrifuge
Supplier: United Product & Instrument
C800-03 C800-04
470012-612PK 86.9 CAD
470012-612 470012-614
Variable Speed Centrifuge
Centrifuges Benchtop Centrifuges
A Valuable Addition to Any Laboratory.
  • 30 minute timer with bell tone signal
  • Latching lid with Safety Switch
  • Easily accessible stainless steel rotor
For a clean, complete separation of cellular components yielding a superior serum or plasma specimen, use this powerful variable speed centrifuge. Suction cup rubber feet for stability also reduce vibration. Centrifuge features all UL approved components.