Fischer Technical Acrylic Vacuum Plate with Rubber Gasket and Shut Off Valve
Supplier: FISCHER TECHNICAL CO SE
PP-001A
470100-526EA 357.05 CAD
470100-526
Fischer Technical Acrylic Vacuum Plate with Rubber Gasket and Shut Off Valve
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Acrylic high vacuum plate with a 27 cm diameter working surface and a shut-off value.
Comes with a black rubber gasket to create a better seal.