Urease
Supplier: ALDON CORP SE
UU0050-25G
UU0050-10G
470303-002EA
75.2
CAD
470303-002
470303-000
Urease
Chemicals for Science Education
CAS Number: 9002-13-5
Solubility: Water
Synonyms: Bean Meal
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...