Universal Indicator Solution
Supplier: ALDON CORP SE
CAS Number: Mixture
Formula: Mixture
Density: 0.794 g/mL
Boiling and Freezing Point: 75-80°C, -114°C
Solubility: Water
Synonyms: Universal pH Indicator
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: This product is flammable.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...