Types of Fossilization Collection
Supplier: SCOTT RESOURCES 2, LLC SE
3010-S
470111-000EA 61.85 CAD
470111-000
Types of Fossilization Collection
Educational Materials Earth Science Educational Materials Paleontology Learning Activities
This Collection has been assembled to provide background into the history of life on Earth.
  • Collection Contains Real Fossils Collected from Multiple Locations
  • All Major Methods of Fossilization Represented
  • Identification Key Included
  • Average Sample Size: 1/2 to 1"
  • Fifteen Fossils Included
Each set contains individually numbered specimens housed in a compartmented collection box with removable tray and key list. The average sample size is 1/2 to 1".
This illustrative set of 15 fossils displays examples of the primary forms of fossilization including direct replacement (pyritization, calcification, silification), molds and casts, distillation (carbonization), and unaltered remains.