High-Vacuum Pumps
Supplier: FISCHER TECHNICAL CO SE
LAV-3/459F LAV-3/G
470200-360EA 2186.7 CAD
470200-360 470200-358
High-Vacuum Pumps
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
High-Vacuum Pumps are ideal for demonstrations and lab work.
  • Intake Connections Via 3/8" ID Tubing
  • Pressure Low Enough for Boyle's Law Experiments
  • Version with Gauge Reduces Errors Due to Poor Seals
  • CSA Approved
  • Fitting for Demos and Lab Work
Intake connections are via 3/8" ID tubing. Both models deliver pressure low enough for all Boyle's Law experiments.
The version with gauge reduces errors due to a poor seal between the pump and the vacuum plate.
Airflow: 3 cfm
Max vacuum: 25 microns (0.025 mm of Hg)
120 V
Certifications: CSA approved.