Twistit® Stopper Assortment
Supplier: PLASTICOID CO INC THE SE
ZM299ASST-B
470151-194CS 1161.7 CAD
470151-194
Twistit® Stopper Assortment
Stoppers Rubber Stoppers
Solid, One-Hole, Two-Hole, and Three-Hole Rubber Stoppers in One Unit.
  • Easily converts from solid to up to 3-hole stopper
  • Available in sizes 3 to 10
  • 1 lb. bag containing 33 stoppers
Consists of nine size 3 stoppers, eight size 4, six size 5, four size 6, two size 7, two size 8, one size 9 and one size 10 stopper. Sizes 3 and 4 stoppers have 2 holes maximum.
Certifications: CAUTION: Item contains latex, which is known to cause allergic reactions in some people. Use with caution.