Turbidity
Supplier: LA MOTTE CHEMICAL PROD CO SE
7519-01 7520-H
470203-128KT 177.75 CAD
470203-128 470002-188
Turbidity
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab. 
  • Range: Five to 200 JTU
  • Material for 50 Tests
  • Compact, Waterproof Carrying Case Contains Kit
  • Instructions Included
  • Pre-calibrated Test Tubes
A compact, waterproof carrying case contains reagents, labware, and step-by-step instructions.
The tests have a range of 5 to 200 JTU.