Trundle Wheel
Supplier: SI Manufacturing
45550
470110-724EA 49.1 CAD
470110-724
Trundle Wheel
Measuring and Leveling Tools
A Fast and Simple Way to Measure Long Distances.
  • Wheel circumference is 1 m
  • Produces audible click with every revolution for tracking purposes
  • 60 cm long handle