Tripod Stand and Accessory Kit
Supplier: GLOBAL MARKET CONNECTIONS, LTD
AK2001
470012-324CS 500.4 CAD
470012-324
Tripod Stand and Accessory Kit
Tripods
This kit is for use with the Laboratory Micro Burner.
  • Three Locking Grooves
  • Set of Three Clay Triangles
Lengthen the stand to 9" with a set of three legs.
Ordering information: Stand, Tripod Legs, keys and Accessory Kit available separately. The Accessory Kit includes a wire gauze pad, evaporating dish holder, and a set of three clay triangles for 60, 70, and 80 mm crucibles.