Ward's® Live Green Tree Frog (Hyla sp.)
Supplier: Avantor
878190
470176-342PK 364.35 CAD
470176-342 470180-580
Ward's® Live Green Tree Frog (Hyla sp.)
Specimens Live Specimens Animals - Living Material
These fascinating frogs are uniquely adapted for climbing, with prominent adhesive pads on the ends of their digits.
  • Makes for a great lab or classroom pet.
  • Will occasionally vocalize, especially during springtime.
Keep in glass or plastic terrarium. Will feed on small insects such as cricket nymphs or flightless fruit flies.
Ordering Information: Care instructions for these frogs are available online.
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd dayMore Product Information