Tree Calipers
Supplier: PECO SALES INC SE
59722
470021-026EA 604.36 CAD
470021-026
Tree Calipers
Calipers
Easily Take Accurate Measurements in the Field
Made of aluminum, this lightweight tree caliper is a valuable forestry tool designed for ease of use. Measure the diameter of tree trunks and other large objects up to 24 inches with the precision of 1/10 inch graduations marked on both sides. Size: 34" x 16".