Transformation of Escherichia coli Lab, Edvotek
Supplier: Edvotek
221 221
470193-134EA 209.5 CAD
470193-134 470200-568
Transformation of Escherichia coli Lab, Edvotek
Educational Classroom Kits and Activities
Understand the Basics of Genetic Engineering
  • Guided or open-inquiry
  • Bactobeads are stable for nine months at room temp
  • Time required: ~90 minutes over 2-3 days
In this experiment, your students will develop an understanding of bacterial transformation by plasmid DNA by introducing an opportunity to observe an acquired phenotypic trait of the transformed bacterial cells. The presence of blue bacterial colonies visually demonstrates the expression of a specific gene for the Lac+ phenotype.
Kit includes: instructions, BactoBeads™, plasmid DNA, buffer, media, ampicillin, X-Gal, ReadyPour™ agar, petri dishes, sterile pipettes, loops and microtubes.
Required, but not included: micropipette and tips, two water baths (37˚C and 42˚C), incubation oven, thermometer, pipette pumps or bulbs, ice, marking pens, Bunsen burner, hot plate or microwave oven, and hot gloves
Packaging: Some components require refrigerator & freezer storage on arrival.