Toluidine Blue O
Supplier: ALDON CORP SE
CAS Number: 92-31-9
Formula: C15H16CIN3S
Density: 1.008 g/mL
Solubility: Water and Alcohol
Synonyms: Basic Blue 17 and Tolonium Chloride, C.I. No 52040
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...