Toluene

Supplier: ALDON CORP SE

TT0190-1LR TT0190-500MLR
470302-948EA 48.8 CAD
470302-948 470302-950
Toluene
Chemicals for Science Education

CAS Number: 108-88-3
Formula Weight: 92.14
Formula: C6H5CH3
Density (g/mL): 0.863
Boiling Point (°C): 109-112
Freezing Point (°C): -95
Solubility: Alcohol
Synonyms: Toluol, Methyl Benzene
Shelf Life (months): 36
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR