Tissue Forceps With Teeth
Supplier: United Scientific Supplies
FOBL45 141015 141017
470014-712EA 12.15 CAD
470014-712 470018-964 470018-966
Tissue Forceps With Teeth
Forceps
A true no slip hold.
  • Interlocking teeth prevent slipping
  • Ridged handles for a confident hold
  • Stainless construction prevents chipping
For general dissection, these 1 × 2 teeth firmly grasp tissues, while the serrated handles provide nonslip control. Gillie's type with straight blunt tips.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.