Tirrill Burners
Supplier: Shiv Dial Sud & Sons
C-1480/3 C-1480/1
470148-642CS 581.5 CAD
470148-642 470148-694
Tirrill Burners
Heaters Gas Burners
Precise Flame Control.
  • Gas Inlet: 3/8" O.D.
  • Mixing Tube: 1/2" O.D.
  • Height: 51/4"
  • LP gas burner, compatible with propane
These burners allow you to adjust both the gas and air supply for precise flame control. The aluminum mixing tube features a flame stabilizer, as well as both needle valve and air ports. It has a nickel-plated zinc base.