Tin (II) Chloride Dihydrate

Supplier: ALDON CORP SE

SS0938-CONC SS0937-100G SS0937-500G SS0937-100GR SS0939-500ML
470302-920EA 11.45 CAD
470302-920 470302-922 470302-924 470302-926 470225-812
Tin (II) Chloride Dihydrate
Chemicals for Science Education

CAS Number: 10025-69-1
Formula Weight: 225.63
Formula: SnCl2·2H2O
Density (g/mL): 2.71
Freezing Point (°C): 38
Solubility: Water, Alkalies, Tartaric Acid and Alcohol
Synonyms: Stannous Chloride, Dihydrate
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR