Tin

Supplier: ALDON CORP SE

TT0175-100G TT0170-100GG TT0170-500GG TT0170-100GR TT0180-100G TT0170-500G
470302-942EA 62.25 CAD
470302-942 470302-934 470302-936 470302-938 470302-944 470302-946
Tin
Chemicals for Science Education

CAS Number: 7440-31-5
Formula Weight: 118.69
Formula: Sn
Density (g/mL): 7.31
Boiling Point (°C): 231.9
Freezing Point (°C): 2270
Solubility: Acids, Potassium Hydroxide Solution
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR