Thymolphthalein
Supplier: ALDON CORP SE
CAS Number: 125-20-2
Formula Weight: Mixture
Formula: Mixture
Density (g/mL): 0.79
Boiling Point (°C): 74-80
Freezing Point (°C): -114
Solubility: Alcohol and Acetone
Shelf Life (months): 36
Storage: Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...