Thionin

Supplier: ALDON CORP SE

TT0105-1G TT0106-100ML
470302-902EA 30.9 CAD
470302-902 470225-690
Thionin
Chemicals for Science Education

CAS Number: 581-64-6
Formula Weight: 263.75
Formula: C12H10N3SCl
Solubility: Hot Water
Synonyms: Lauth's Violet, Phenothiazin-5-ium, 3,7-diamino-, chloride
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR