Thermostat
Supplier: VWR International
PH0403
470005-892EA 42.5 CAD
470005-892
Thermostat
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
This working model thermostat will demonstrate how differential expansion is used to control temperature. A bimetallic strip is used to open or close an electric circuit when temperature changes. The contacts will carry about three amperes. The unit can be operated with a 6 V lantern battery, low voltage power supply, or a blow dryer. Mounted on a metal base; approximately 15 cm high.