The Water Quality Educator and Monitoring Outfit
Supplier: LaMotte
5870-01 R-5860 R-3121-01 7520-H R-3354
470004-822KT 886.5 CAD
470004-822 470112-688 470199-990 470199-994 470199-996
The Water Quality Educator and Monitoring Outfit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
This comprehensive kit includes everything needed to develop a curriculum and conduct studies in monitoring water quality.
  • Seven individual test kits
  • Each test can be repeated 50 times
  • Handbook and interactive CD-ROM
  • Aligned to NGSS MS-ESS3-3, HS-LS2-7, HS-ESS2-5, HS-ESS3-4
Comprised of two components, The Water Quality Educator and The Water Quality Monitoring Outfit, this kit is available as a complete set or individually. The Water Quality Educator contains an interactive CD-ROM (PC and MAC compatible) and a 64 page Monitor’s Handbook that provide substantial classroom, laboratory, and field support for testing procedures and activities.
The Water Quality Monitoring Outfit contains seven individual test kits for testing pH, Dissolved Oxygen, Nitrate, Alkalinity, Phosphate, Turbidity, and Temperature. Each test can be repeated 50 times, excluding pH and temperature, which can be tested 100 times and unlimited respectively. Individual test kit reagent refills are also available.
Ordering information: This kit includes material for 50 tests.