The Ultrastructure of Animal Cells Microslide
Supplier: SUPERTEK SCIENTIFIC LLC
T-66 T-66-15B
470009-036PK 19.6 CAD
470009-036 470022-116
The Ultrastructure of Animal Cells Microslide
Slides Prepared Slides
Investigate cellular components up close!
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Gland cells (cs, st, 400X), Plasma cell (cs, st, 37,000X), Nerve cell membrane (cs, st, 400,000X), Golgi body and vacuole (cs, st, 26,000X), Mitochondrion (cs, st, 124,000X), Centriole (cs, st, 175,000X), Lamp-brush chromosomes (cs, st, 700X), and Muscle cell fibrils (cs, st, 45,000X).