Tetrahymena Medium
Supplier: Avantor
WARD470180-756
470180-756PK 59.99 CAD
470180-756
Tetrahymena Medium
Cell Culture Media
Sterile. For culture of Tetrahymena. Supplied in 20 x 150 mm tubes. Package of 12.