Swift M3700 + M3800 Series Advanced Microscopes
Supplier: Swift Instruments
M3802CT-4 M3802CB-4
470044-070EA 952.05 CAD
470044-070 470308-158 470308-160
Swift M3700 + M3800 Series Advanced Microscopes
Microscopes
Packed with standard research features, the Swift M3700 + M3800 series advanced biological microscopes are ideal for educational and professional applications.
  • Viewing head is inclined 45° and rotatable 360° for ergonomic viewing
  • Objectives: 4×, 10×, 40×, 100×
  • Built-in mechanical stage
  • Energy-efficient, cordless LED illumination
Combining a new design with a built-in handle that encourages proper handling, this microscope fits the needs of taxing clinical and student environments.
Objectives are secured in a rotatable inward-facing reversedMore Product Information