Cotton-Tipped Swab Applicators
Supplier: PERRIGO DIRECT , INC.
USC00051
470150-658CS 488.8 CAD
470150-658 470019-172
Cotton-Tipped Swab Applicators
Swabs Swabs for Sampling
Variety of sterile or non sterile swabs.
Sterile swabs have wooden shafts and are indivdually sealed. Depending on the pack size ordered, they may be packaged in a bag (pack/10) or box (pack/100). Excellent for spreading a confluent lawn of bacteria without breaking the agar surface.
Non sterile swabs are packaged together in a box or bag. Excellent for manipulating small samples that do not need to be kept sterile.
Ordering information:
For educational purposes only.