Support Stands with Rods and Rings
Supplier: United Scientific Supplies
SET462-SK CH12380/3 SET694-SK
470005-864KT 97.35 CAD
470005-864 470005-866 470104-528
Support Stands with Rods and Rings
Retort Stands
A Convenient Method of Purchasing Both Rings and Supports
  • Sets comes with 2, 3, or 4 rings of varied diameters
  • Base is stamped steel and the plate underneath is made of steel
  • Base weighs 2 lbs.
The support stand will support up to 10 lbs.